Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TYMP Mouse Monoclonal Antibody [clone: OTI1G10]
Anti-TYMP Mouse Monoclonal Antibody [clone: OTI1G10]
Catalog # ORIGTA800961
Supplier:  OriGene
undefined
Anti-TYMP Mouse Monoclonal Antibody [clone: OTI1G10]
Catalog # ORIGTA800961
Supplier:  OriGene

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Thymidine Phosphorylase
  • Symbol antigenu:
    TYMP
  • Klonovalita:
    Monoclonal
  • Klon:
    OTI1G10
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • Izotyp:
    IgG1
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • Formulář:
    Liquid
  • Počet aminokyselin:
    311 to 482
  • Skladovací pufr:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Imunogen:
    Protein
  • Čištění:
    purified by affinity chromatography
  • Bal.:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TYMP (thymidine phosphorylase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human