Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ENTPD2 Rabbit Polyclonal Antibody
Anti-ENTPD2 Rabbit Polyclonal Antibody
Catalog # ORIGTA317967
Supplier:  OriGene
Anti-ENTPD2 Rabbit Polyclonal Antibody
Catalog # ORIGTA317967
Supplier:  OriGene

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    ectonucleoside triphosphate diphosphohydrolase 2
  • Symbol antigenu:
    ENTPD2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    internal
  • Křížová adsorbce:
    none
  • Formulář:
    Liquid
  • Skladovací pufr:
    PBS, 0,1% sodium azide.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by immunoaffinity chromatography
  • Bal.:
    50 µG

Specifications

About this item

Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: ENTPD2 (ectonucleoside triphosphate diphosphohydrolase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human