Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GPR151 Rabbit Polyclonal Antibody
Anti-GPR151 Rabbit Polyclonal Antibody
Catalog # ORIGTA316108
Supplier:  OriGene
Anti-GPR151 Rabbit Polyclonal Antibody
Catalog # ORIGTA316108
Supplier:  OriGene

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    G Protein-Coupled Receptor 151
  • Symbol antigenu:
    GPR151
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Křížová adsorbce:
    none
  • Formulář:
    Liquid
  • Skladovací pufr:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by affinity chromatography
  • Bal.:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: GPR151 (G protein-coupled receptor 151)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human