Anti-RNF138 Rabbit Polyclonal Antibody
ORIGTA315477
: OriGene
- Druh protilátky:Primární
- Název antigenu:Ring Finger Protein 138
- Symbol antigenu:RNF138
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Epitop:internal
- Křížová adsorbce:none
- Formulář:Liquid
- Skladovací pufr:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Čištění:purified by affinity chromatography
- Bal.:100 µl
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000
Type: Primary
Antigen: RNF138 (ring finger protein 138, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat