Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-UBTF Rabbit Polyclonal Antibody
  ORIGTA314660
 :  OriGene
Anti-UBTF Rabbit Polyclonal Antibody
  ORIGTA314660
 :  OriGene

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Upstream binding transcription factor, RNA polymerase I
  • Symbol antigenu:
    UBTF
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    none
  • Formulář:
    Liquid
  • Modifikace:
    phospho-Ser484
  • Skladovací pufr:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by affinity chromatography
  • Bal.:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: UBTF (upstream binding transcription factor, RNA polymerase I)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat