Anti-PPP2R3B Rabbit Polyclonal Antibody
ABGEAP7772A
: Abgent
- Druh protilátky:Primární
- Název antigenu:Protein Phosphatase 2, Regulatory Subunit 3B
- Symbol antigenu:PPP2R3B
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Epitop:N-terminal
- Synonyma antigenu:Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta|Protein phosphatase 2A 48 kDa regulatory subunit|subunit B|NY-REN-8|PPP2R3B|PP2A|PR48 isoform|PPP2R3LY|PPP2R3L
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Bal.:400 µl
Type: Primary
Antigen: PPP2R3B (protein phosphatase 2, regulatory subunit B'', beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human