Anti-CDK5R1 Rabbit Polyclonal Antibody
ABGEAP7743B
: Abgent
- Druh protilátky:Primární
- Název antigenu:Cyclin-dependent kinase 5 regulatory subunit 1
- Symbol antigenu:CDK5R1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:C-Terminal
- Identifikační číslo genu:8851
- Synonyma antigenu:p35nck5a|CDK5R|p35|p23|CDK5P35|NCK5A|p25
- Skladovací pufr:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molekulární hmotnost:34.06 kDa
- Sekvence:CSDLKNESGQEDKKRL
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0,5 mg/ml
- Transportní teplota:4 °C
- Imunogen:This CDK5R1(p35) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 272-301 amino acids from the C-terminal region of human CDK5R1(p35).
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Type: Primary
Antigen: CDK5R1 (cyclin-dependent kinase 5, regulatory subunit 1 (p35))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human