Anti-LRP3 Rabbit Polyclonal Antibody
Catalog # ABGEAP6155A
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:Low density lipoprotein receptor-related protein 3
- Symbol antigenu:LRP3
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:C-Terminal (650–700 aa)
- Křížová adsorbce:No
- Synonyma antigenu:105 kDa low-density lipoprotein receptor-related protein|Low-density lipoprotein receptor-related protein 3|hLRp105|LRP-3|LRP3
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: LRP3 (low density lipoprotein receptor-related protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human