Anti-ZBTB8B Rabbit Polyclonal Antibody
Catalog # ABGEAP16258B
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:Zinc Finger And BTB Domain Containing 8B
- Symbol antigenu:ZBTB8B
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:C-Terminal
- Identifikační číslo genu:728116
- Synonyma antigenu:ZBTB8B|RP1 27O5.1|Zinc finger and BTB domain-containing protein 8B|RP1-27O5.1|ZBT8B_HUMAN|ZNF916B.|Putative zinc finger and BTB domain containing protein 8B
- Skladovací pufr:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molekulární hmotnost:54.175 kDa
- Sekvence:VESGEENDPAGDDSC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0,35 mg/ml
- Transportní teplota:4 °C
- Imunogen:This ZBTB8B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 450-478 amino acids from the C-terminal region of human ZBTB8B.
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: ZBTB8B (zinc finger and BTB domain containing 8B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human