Anti-KIDINS220 Rabbit Polyclonal Antibody
Catalog # ABGEAP16256B
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:kinase D-interacting substrate, 220kDa
- Symbol antigenu:KIDINS220
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:C-Terminal
- Identifikační číslo genu:57498
- Synonyma antigenu:ARMS
- Skladovací pufr:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molekulární hmotnost:196.542 kDa
- Sekvence:NTPLLKDDKDRKAEGC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0,5 mg/ml
- Transportní teplota:4 °C
- Imunogen:This KIDINS220 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1518-1547 amino acids from the C-terminal region of human KIDINS220.
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: KIDINS220 (kinase D-interacting substrate, 220kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human