Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SMEK2 Rabbit Polyclonal Antibody
Anti-SMEK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP16035B
Supplier:  Abgent
Anti-SMEK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP16035B
Supplier:  Abgent

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Protein Phosphatase 4, Regulatory Subunit 3B
  • Symbol antigenu:
    PPP4R3B
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Identifikační číslo genu:
    57223
  • Synonyma antigenu:
    PSY2|FLFL2|PP4R3B|smk1|SMEK2
  • Skladovací pufr:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molekulární hmotnost:
    97.458 kDa
  • Sekvence:
    PKPEDDFPDNYEKFC
  • Skladovací teplota:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Koncentrace:
    0,5 mg/ml
  • Transportní teplota:
    4 °C
  • Imunogen:
    This SMEK2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 720-748 amino acids from the C-terminal region of human SMEK2.
  • Čištění:
    Protein A & Peptide affinity purification
  • Bal.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SMEK2 (SMEK homolog 2, suppressor of mek1 (Dictyostelium))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human