Anti-RAB11A Rabbit Polyclonal Antibody
Catalog # ABGEAP14530C
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:RAB11A, member RAS oncogene family
- Symbol antigenu:RAB11A
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:Center
- Identifikační číslo genu:8766
- Synonyma antigenu:RAB11A|MGC1490|YL8|Ras-related protein Rab-11A|RAB 11A|member oncogene family|member RAS oncogene family
- Skladovací pufr:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molekulární hmotnost:24.394 kDa
- Sekvence:ERWLKELRDHADSNC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0,4 mg/ml
- Transportní teplota:4 °C
- Imunogen:This RAB11A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 88-116 amino acids from the Central region of human RAB11A.
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: RAB11A (RAB11A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human