Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ALKBH4 Rabbit Polyclonal Antibody
Anti-ALKBH4 Rabbit Polyclonal Antibody
Catalog # ABGEAP12774C
Supplier:  Abgent
Anti-ALKBH4 Rabbit Polyclonal Antibody
Catalog # ABGEAP12774C
Supplier:  Abgent

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    AlkB Homolog 4, Alpha-Ketoglutarate-Dependent Dioxygenase
  • Symbol antigenu:
    ALKBH4
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    middle
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Bal.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ALKBH4 (alkB, alkylation repair homolog 4 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse