Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PNLDC1 Rabbit Polyclonal Antibody [clone: 217CT22.1.4]
Anti-PNLDC1 Rabbit Polyclonal Antibody [clone: 217CT22.1.4]
Catalog # ABGEAP11385B
Supplier:  Abgent
Anti-PNLDC1 Rabbit Polyclonal Antibody [clone: 217CT22.1.4]
Catalog # ABGEAP11385B
Supplier:  Abgent

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Poly(A)-specific ribonuclease (PARN)-like domain containing 1
  • Symbol antigenu:
    PNLDC1
  • Klonovalita:
    Polyclonal
  • Klon:
    217CT22.1.4
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Synonyma antigenu:
    SRS|MRSR|spermidine aminopropyltransferase|Spermine synthase|SPMSY|SpS
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Bal.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PNLDC1 (poly(A)-specific ribonuclease (PARN)-like domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human