Anti-MYBPC3 Rabbit Polyclonal Antibody
Catalog # AVIVARP41377_P050
Supplier: Aviva Systems Biology
Specifications
- Druh protilátky:Primární
- Název antigenu:Myosin binding protein C, cardiac
- Symbol antigenu:MYBPC3
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Epitop:C-terminal
- Křížová adsorbce:none
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Bal.:50 µG
Specifications
About this item
Type: Primary
Antigen: MYBPC3 (myosin binding protein C, cardiac)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human