Anti-RORC Rabbit Polyclonal Antibody
Catalog # AVIVARP37127_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: RORC (RAR-related orphan receptor C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Rat
Specifications
- Western blot:Yes
- Hostitel:Rabbit
- Klonovalita:Polyclonal
- Synonyma antigenu:Nr1f3|Thor|TOR|RORgamma
- Identifikační číslo genu:19885
- Druh protilátky:Primární
- Epitop:N-terminal
- Symbol antigenu:RORC
- Cat. No.:AVIVARP37127_P050
- Reaktivita:Human|Guinea pig|Dog|Rat|Mouse
- Transportní teplota:Ice
- Křížová adsorbce:none
- Skladovací teplota:Cool
- Spojení:Unconjugated
- Název antigenu:RAR-related orphan receptor gamma
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



