Cap strips for skirted low profile plates
: Brand
Frequently Bought Together
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Latexové rukavice
: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
: VWR Collection
VWR®, Ubrousky čisticí
: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Real-Time PCR Plates, 96-Well and 384-Well
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
PCR destičky, 96-jamek
: Brand