Carboys, space saver, with screw cap, SP Bel-Art
Catalog # BELAH109390005
Supplier: Bel-Art Products, a Part of SP
Specifications
- Kapacita:5 l
- Š×H×V:241×89×375 mm
- Bal.:1
Specifications
About this item
PP, translucent.
- In addition to the top handle, a convenient grip hold in the front facilitates removal from a shelf
- Graduated markings on front in liters
- Good chemical and temperature resistance
Space saver book bottle stacks book-like on shelves for efficient use of space.