ContactUsLinkComponent
Containers for plant tissue culture
Containers for plant tissue culture
Katalogové číslo 212-3219
Dodavatel:  Greiner Bio-One
undefined
Containers for plant tissue culture
Katalogové číslo 212-3219
Dodavatel:  Greiner Bio-One
Specifikace
  • Ø:
    53 mm
  • Balené:
    Bulk
  • Kapacita:
    175 ml
  • Popis:
    Bottom part, without lid, non sterile
  • Výška:
    100 mm
  • Sekvence:
    03
  • Bal.:
    315
Specifikace
O tomto produktu
Container, PS, clear for plant tissue culture. These containers are specially suited for plant tissue culture. The use of a very clear material ensures maximum light transmission and thus rapid and successful growth. These culture containers are not only suitable for the proliferation of plant tissue cultures but can also be used as transport containers.
  • Available with or without lid
  • Polystyrene for high clarity
  • ith flat bottom
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904