Labtop coolers, LabCooler™
REVC6744
- Popis:LabCooler™ II, benchtop
- Environmentally Preferable:
- Bal.:1
Benchtop cooler −20 °C.
It helps maintain enzymes and other frozen material at stable temperatures when used at the lab bench. When stored in a freezer, coolers insulate enzymes and other biologicals from periodic temperature fluctuations due to door openings, defrost cycles or temporary power outages of short duration.
It maintains enzymes at stable −20 °C C for up to two hours. Holds (24) 1,5 ml tubes and (8) 0,5 ml tubes.