Deoxyribonuclease I
0,5 KU
Katalogové číslo ROCKMB-101-0500
Dodavatel: Rockland Immunochemicals
Přejít na:
Product Details & Documents
Deoxyribonuclease I (usually called DNase I), is an endonuclease coded by the human gene DNASE1. DNase I is a nuclease that cleaves DNA preferentially at phosphodiester linkages adjacent to a pyrimidine nucleotide, yielding 5'-phosphate-terminated polynucleotides with a free hydroxyl group on position 3', on average producing tetranucleotides. It acts on single-stranded DNA, double-stranded DNA, and chromatin.
Specifikace
Specifikace
- Bal.:0,5 KU
- Cat. No.:ROCKMB-101-0500
- Třída:Molecular biology grade
- Název enzymu:Deoxyribonuclease I