Orders LinkContactUsLinkComponent
Capillary columns, HP-5ms
 :   Agilent
Capillary columns, HP-5ms
 :  Agilent
Capillary columns, HP-5ms
Capillary columns, HP-5ms
These columns have very low bleed characteristics that are ideal for GC/MS.
  • Specifically tested for inertness for active compounds including acidic and basic compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded and cross-linked
5%-Phenyl-methylpolysiloxane, non-polar, solvent rinsable
Product Family Options
  • Sekvence
    Popis
    Formát kolony
    Fáze
    Tloušťka filmu
    Ø vnitř.
    Délka
  • 02
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,18 µm
    0,18 mm
    20 m
    1 * 1 KS
    1 * 1 KS
  • 04
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,33 µm
    0,20 mm
    12 m
    1 * 1 KS
    1 * 1 KS
  • 07
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,33 µm
    0,20 mm
    25 m
    1 * 1 KS
    1 * 1 KS
  • 10
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,10 µm
    0,25 mm
    15 m
    1 * 1 KS
    1 * 1 KS
  • 11
    on 7" cage
    Capillary
    HP-5ms
    0,25 µm
    0,25 mm
    15 m
    1 * 1 KS
    1 * 1 KS
  • 12
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,25 µm
    0,25 mm
    15 m
    1 * 1 KS
    1 * 1 KS
  • 18
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,25 µm
    0,25 mm
    30 m
    1 * 1 KS
    1 * 1 KS
  • 19
    on 5975T LTM toroid
    Capillary
    HP-5ms
    0,25 µm
    0,25 mm
    30 m
    1 * 1 KS
    1 * 1 KS
  • 30
    on 7890/6890 LTM module
    Capillary
    HP-5ms
    0,25 µm
    0,32 mm
    30 m