Capillary columns, DB-17ms
Catalog # HEWL122-4732
Supplier: Agilent
Specifications
- Formát kolony:Capillary
- Tloušťka filmu:0,25 µm
- Ø vnitř.:0,25 mm
- Popis:on 7" cage
- Délka:30 m
- Sekvence:05
- Fáze:DB-17ms
- Bal.:1
Specifications
About this item
Virtually equivalent to (50%-Phenyl)-methylpolysiloxane column. They are solvent rinsable and very low bleed mid-polarity columns ideal for GC/MS. They are suitable for CLP pesticides.
- Excellent inertness for active compounds
- Enhanced mass spectral integrity
- Bonded and cross-linked
Max. temperature: 40 to 320/340 °C