Phosphorus (Reactive) TNT reagent set
Catalog # 705-2049
Supplier: Hach
CAS Number:
Specifications
- Chemická metoda:Molybdovanadate
- Bal.:1 SET
- Popis:Phosphorus TNT reagent set, High range
- Pro jednotku:DR900
- Testů v kitu:50
- Rozsah:1,0 - 100,0 mg/L PO₄
Specifications
About this item
Set used for determination of reactive phosphorus (ortho-phosphate) by the molybdovanadate Test 'N Tube™ method.
- Test 'N Tube products provide safe, convenient testing
- Easy ordering
Delivery: Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).