Visual colour kits, phosphorus, PO-19A
Catalog # HACH224801
Supplier: Hach
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Phosphorus, orthophosphate test kit, model PO-19A
- Rozsah:0 - 0,8 mg/l PO₄
0 - 4 mg/l PO₄
0 - 40 mg/l PO₄
Specifications
About this item
Simple, economical rapid colour disc method with an easy to carry test kit.
Delivery: Kit includes two reagents, colour disc, comparator box, two viewing tubes, viewing adaptor, two bottles, dropper tube, funnel, filter paper, deionized water, instructions and carrying case.