Multi parameter test kit
HACH223002
: Hach
:
- Bal.:1 KIT
- Popis:Multi parameter test kit, CN-39WR
This multi-test kit combines popular water treatment tests into one compact kit which can be used to determine chlorine (0 - 3,4 mg/l), hardness (1 - 20 gpg), iron (0 - 4 mg/l) and pH (4 - 10) in the water sample.
- Includes frequently measured water quality parameters Uses simple, economical methods
Hardeness testing: gpg= grains per gallon
: Includes six reagents, bottles, colour discs, comparator box, viewing tubes, measuring tube, instructions and carrying case.