ContactUsLinkComponent
Turbidity standard, ULR verification kit, Stablcal®
Turbidity standard, ULR verification kit, Stablcal®
Catalog # HACH2714600
Supplier:  Hach
CAS Number:  
undefined
Turbidity standard, ULR verification kit, Stablcal®
Catalog # HACH2714600
Supplier:  Hach
CAS Number:  
Specifications
  • Bal.:
    1 KIT
  • Měření zákalu:
    1,0 NTU
    0,30 NTU
    0,10 NTU
    0,50 NTU
  • Popis:
    ULR verification kit, Stablcal®
  • Pk Info:
    4x 100 ml
Specifications
About this item
StablCal verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 mL each), sililcone oil, oiling cloth and instructions is used for verification purpose only.
  • Precise, pre-mixed formazin solutions Enhanced stability saves time and labor Confidence in results