Mobile Storage Systems, KP Type PLUS
720-2816
- Popis:KP-GLASS PLUS, mobile storage system, metal cabinet, 11 rows with 26 drawers each, blue - capacity: 121000 slides
- Sekvence:02
- Š×H×V:1000×500×1800 mm
- Bal.:1
For the storage of slides and cassettes.