Acidity reagent set for digital titrator
Catalog # HACH2272800
Supplier: Hach
CAS Number:
Specifications
- Bal.:100 Tests
- Popis:Acidity reagent set, 100 tests
Specifications
About this item
This set is used for the determination of acidity by digital titrator titration with methyl orange and phenolpthalein end points.
- Fast and Flexible
- Versatile
- Economical
- Accurate
Reagent Set includes: Bromphenol blue powder pillows, phenolphthalein powder pillows and two NaOH cartridges (0.1600 N and 1.600N). Ranges: 10 to 160 and 1000 to 4000 mg/L as CaCO₃.