Visual colour comparator kit, phosphate
Katalogové číslo HACH224902
Dodavatel: Hach
Číslo CAS:
Specifikace
- Bal.:1 KIT
- Popis:Phosphate test kit, (ortho+meta), model PO-23
- Rozsah:0,1 - 4 mg/l
1 - 40 mg/l
Specifikace
O tomto produktu
Simple, economical rapid colour disc method with an easy to carry test kit.
Delivery: Kit includes three reagents, colour disc, comparator box, two viewing tubes, bottle, dropper tube, stove and cover, test tube clamp, deionised water, instructions and carrying case.