Visual comparator test kits, nitrate-nitrite, NI-12
Catalog # HACH1408100
Supplier: Hach
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Nitrate - Nitrite test kit, model NI-12
- Typ:Cadmium reduction, diazotisation
- Rozsah:0 - 40 mg/l NO₃-N, 0.01 - 0.40 mg/l NO₂- N
Specifications
About this item
Simple, low cost colour disc method of testing nitrate-nitrite.
Delivery: Includes colour disc, comparator box and tubes, reagents, instructions and carrying case.