Aluminium reagent set for Al determination by photometric ECR method
HACH2443400
: Hach
:
- Chemická metoda:Eriochrome cyanine R
- Bal.:1 SET
- Popis:Aluminium reagent set, 100 tests
- Pro jednotku:DR2000, DR3000
- Rozsah:0,002 - 0,220 mg/l
Kit used for the determination of aluminium by Eriochrome Cyanine R (ECR) method and has sample size of 50 ml.
: Set contains ECR reagent powder pillows, hexamethylenetetramine buffer reagent powder pillows and ECR masking reagent solution for approximately 100 tests.