Orders LinkContactUsLinkComponent
Immersion coolers, RU series, Techne®
Katalogové číslo
Immersion coolers, RU series, Techne®
Katalogové číslo
Dodavatel:  
Immersion coolers, RU series, Techne®
Product Details & Documents
Techne® Dip Coolers are designed for use with Techne liquid baths when temperatures down to –35 °C are required. The cooling head of the Dip Cooler fits neatly and unobtrusively into the corner of the bath and can be secured with a specially designed mounting bracket (supplied). If cooling can be achieved by using cold tap water, a dip cooler is recommended as it conserves water and is easier and more convenient to use.
  • Choice of two models
  • Compact refrigeration units for achieving temperatures down to –35 °C
  • Cooling head of the Dip Cooler fits neatly into the corner of a Techne liquid bath (RU series of Dip Coolers)
 : Note: Cooling capacity at +20, 0 and −10 °C.