Order Entry
Czech Republic
ContactUsLinkComponent
Spare desiccator lid, PYREX®
Spare desiccator lid, PYREX®
Catalog # CORN3121-250CO
Supplier:  Corning
   Waste Reducing
undefined
Spare desiccator lid, PYREX®
Catalog # CORN3121-250CO
Supplier:  Corning
   Waste Reducing

Specifications

  • Nominální velikost:
    250
  • Type:
    PYREX® NS 24/29
  • Popis:
    Replacement cover for PYREX® 10.5 L large top desiccator (for use with 24/29 stopcock)
  • Environmentally Preferable:
    Waste Reducing
  • Bal.:
    1

Specifications

About this item

PYREX® replacement cover for 10,5 L large top desiccator using 24/29 Standard Taper stopcock.

Ordering information: Stopcock not included.