Příslušenství pro vakuové filtrační systémy Nalgene®
Catalog # 513-3971
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Filter funnel adapter
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Bal.:25
Specifications
About this item
The non sterile PP adaptor is the bulk-packed version of funnel adapters included with 145- and 147-series analytical test filter funnels. Used in high throughput QC testing laboratories.