Adhesion slides, HistoBond®
Catalog # MARI0810701
Supplier: MARIENFELD
Specifications
- Barva:Bílá
- Popis:Slides with frosted ends
- Bal.:2.000
Specifications
About this item
HistoBond slides combine frosted ends with printed in various colours.
- Soda lime glass of hydrolytic class 3
- Different colours for colour coding of methods of analysis
- Thin-coated marking area facilitates the processing on automated systems
Certifications: IVD directive 98/79/EC