Measuring cylinders, tall form, class A, borosilicate glass, PYREX® VISTA™
Catalog # CORN70022-100
Supplier: Corning
Specifications
- Kapacita:100 ml
- Sekvence:05
- Bal.:12
Specifications
About this item
PYREX® borosilicate glass, class A.
- Calibrated to contain (TC) in accordance with ASTM E-1272
- Hexagonal base with graduations in blue enamel
- 10 ml cylinder has a funnel top