Adenine deaminase
Catalog # 786-428
Supplier: G-Biosciences
CAS Number:
Specifications
- Bal.:0,25 mL
- Popis:Adenine deaminase
Specifications
About this item
Adenine deaminase converts adenine to hypoxanthine. It is a highly purified recombinant enzyme protein supplied at a 1 µM concentration.