Apparatus for the determination of aromatic oils by steam distillation according to DAB 7
Catalog # NEUB2410-10-100
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Velikost objímky:29/32 NS
- Type:Complete set
- Popis:with PTFE stopcock and round-bottom flask, 1000 ml
- Bal.:1
Specifications
About this item
Steam distillation apparatus designed to extract and determine the resulting volume aromatic oils. Apparatus has graduated receiver for organic matter for 10 ml volume in 0,1 ml increments.