Adenosylhomocysteine nucleosidase
786-427
:
- Bal.:0,5 mL
- Název enzymu:Adenosylhomocysteine nucleosidase
This recombinant adenosylhomocysteine nucleosidase (10 µM) converts S-adenosylhomocysteine to S-(deoxy-D-ribos-5-yl)-L-homocysteine and adenine.