Orders LinkContactUsLinkComponent
Flow through cell
Flow through cell
02, Flow through cell, glass, for electrodes with 12 mm diameter
Catalog # METT51302257
Supplier:   Mettler - Toledo
Flow through cell
Catalog # METT51302257
Supplier:  Mettler - Toledo
Flow through cell
02, Flow through cell, glass, for electrodes with 12 mm diameter
Price
9 380,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Documents
Specifications
  • Popis:
    Flow through cell, glass, for electrodes with 12 mm diameter
  • Sekvence:
    02