Fluorescent β-Galactosidase assay kit
Catalog # 786-651
Supplier: G-Biosciences
CAS Number:
Specifications
- Bal.:500 Assays
- Popis:β-Galactosidase Assay Kit
- Testů v kitu:500 assays
Specifications
About this item
β-Galactosidase assay is for determining the β-galactosidase activity in the lysates of cells transfected with a β-galactosidase expression construct. It uses chlorophenol red-β-D-galactopyranoside (CPRG), which is up to 10 times more sensitive than the classic O-nitrophenyl-β-D-galactopyranoside (ONPG) β-Galactosidase assays. This increased sensitivity makes it simpler to measure β-galactosidase activity in cells that are difficult to transfect or have low β-galactosidase activity expression. β-Galactosidase activity is measured using a spectrophotometer or a microplate reader to determine the amount of substrate converted at 570 to 595 nm.
- Colorimetric assay
- 10X more sensitive than ONPG assays
- Scalable
- Monitor levels of transfection in cells