Gas burette, Hempel, Class A
Katalogové číslo WITG2.972.000
Dodavatel: witeg Labortechnik
Specifikace
- Kapacita:100 ml
- Dělení:0,2 ml
- Bal.:1
Specifikace
O tomto produktu
Gas burette, accuracy Class A.
- With resistant DIFFICO graduation
- Main point ring graduation
- Iron base