Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zkumavky
Mikrozkumavky
Accessories for cluster tube and storage racks, 2 ml, BioTube™
Accessories for cluster tube and storage racks, 2 ml, BioTube™
Accessories for cluster tube and storage racks, 2 ml, BioTube™
Catalog # SIMPT105-26
Supplier:
Simport Scientific
Expand image
Accessories for cluster tube and storage racks, 2 ml, BioTube™
Catalog # SIMPT105-26
Supplier:
Simport Scientific
Specifications
Popis:
Mat cover for storage racks with tubes Bio Tube™, non sterile
Bal.:
10
Specifications
About this item
Product Details
Product Details