Magnetic stirring bars positioner/retrievers, Spinbar®, SP Bel-Art
Katalogové číslo BELAH377800000
Dodavatel: Bel-Art Products, a Part of SP
Specifikace
- Ø:15,9 mm
- Length:305 mm
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
PE, to retrieve stirbars without coming into contact with the liquid in the containers.
- No direct contact with stirring magnet
- Splash-free operation