Silicone inserts for pipette containers
Katalogové číslo SCUT3523213
Dodavatel: schuett-biotec GmbH
Specifikace
- Popis:Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
Silicone rubber for insertion in VARICON RS (lower part) and SPECICON (lower and upper part) pipette containers and it is designed to prevent damage to pipettes caused by sudden shock.
Thickness: 2 mm
Ø: 58 mm