Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Jehly ke stříkačkám
Accessories for needle assemblies
Accessories for needle assemblies
HEWLG1329-87017
:
Agilent
Accessories for needle assemblies
HEWLG1329-87017
:
Agilent
Určeno pro:
1220 Infinity LC
Popis:
Seat assembly, 0,17 mm
Sekvence:
23
Bal.:
1