SPE plates, 96-well, HyperSep™, SAX
Katalogové číslo HYPE60300-567
Dodavatel: Thermo Fisher Scientific
Specifikace
- Popis:96-well plate, HyperSep™ SAX
- Sekvence:07
- Náplň:50 mg
- Bal.:1
Specifikace
O tomto produktu
Thermo Scientific™ HyperSep SAX Strong Anion Exchanger SPE 96-Well Plates and Individual Wells feature a strong anion exchange sorbent for extraction of weak acids. The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.