Orders LinkContactUsLinkComponent
Tank tool
Dodavatel:   MICRONOVA
Tank tool
Dodavatel:  MICRONOVA
Certifikáty
Tank tool
4 Dostupných variant níže
Product Details & Documents
The tank tool is designed to efficiently clean chambers, tanks and pipes from above or below. The articulated head allows curved frame to hug the outer wall of the tank as it moves downward to remove residue or apply disinfectant. The unit has a degree of swivel to ensure good contact with the surface being cleaned.
  • Stainless steel frame adapter with swivel action
  • Curved frame maintains contact while cleaning tanks
  • Available in exterior and interior (reverse) configurations
  • Designed for cleanroom use
Frame adapters hold MZSM series of replaceable mop heads. Covers available in a variety of fabrics.
Attaches directly to SSU series handles. Attaches to STAH and STAEH series via SSA-2 Adapter.
Product Family Options
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • 129-1072
    Reverse tank tool adapter, electropolished stainless steel
    Reverse tank tool adapter, electropolished stainless steel
    01
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 070,00 Kč
    10 070,00 Kč
    1 * 1 KS
       10 070,00 Kč
  • 129-1073
    Tank tool adapter, electropolished stainless steel
    Tank tool adapter, electropolished stainless steel
    02
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 070,00 Kč
    10 070,00 Kč
    1 * 1 KS
       10 070,00 Kč
  • MICRQPSLTT-14P
    TankTool™ Exterior Curve, threaded handle connector
    TankTool™ Exterior Curve, threaded handle connector
    03
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 9 430,00 Kč
    9 430,00 Kč
    1 * 1 KS
       9 430,00 Kč
  • MICRQPSLRTT-14B
    TankTool™ Interior Curve, vertical threaded handle connector
    TankTool™ Interior Curve, vertical threaded handle connector
    04
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 3 710,00 Kč
    3 710,00 Kč
    1 * 1 KS
       3 710,00 Kč
Microfibre cover
Informace o produktu
  • Katalogové číslo
    Délka
    Popis
    Sekvence
    Dostupnost
    Price
  • 360 mm
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    22
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 628,00 Kč
    628,00 Kč
    1 * 1 KS
       628,00 Kč