Flow indicator paddle wheel
Flow indicator paddle wheel, SAN, 2 bar
Katalogové číslo 618-1130
Dodavatel: SEMADENI
Přejít na:
Product Details & Documents
Flow indicator with propeller, Styrolacrylnitrile (SAN) transparent.
- Corrosion resistant design without metal parts
- Operating temperature up to 30 °C
- Mounting length 90 mm
Suitable for gas and water flow control.
Upozornení: Not suitable for use with solvents, acids or alkalis.
Specifikace
- Popis:Flow indicator paddle wheel
- Materiál:SAN
- Tlak:2 bar